missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzeneboronic acid, 97%

Catalog No. AAB2434303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2434303 1 g
AAB2434306 5 g
AAB2434314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2434303 Supplier Thermo Scientific Chemicals Supplier No. B2434303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1423-27-4 | C7H6BF3O2 | 189.928 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1423-27-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.928
MDL Number MFCD00236059
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol, o-trifluoromethylphenylboronic acid, 2-boronobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolueneboronic acid
PubChem CID 2734387
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 2974465
Formula Weight 189.93
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only