missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)aniline, 98%

Catalog No. p-7022347
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
AAA1507222 100 g
1 options

Catalog No. AAA1507222

Supplier: Thermo Scientific Chemicals A1507222

Only null left

CAS: 88-17-5 | C7H6F3N | 161.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-17-5
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007718
InChI Key VBLXCTYLWZJBKA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
PubChem CID 6922
IUPAC Name 2-(trifluoromethyl)aniline
SMILES C1=CC=C(C(=C1)C(F)(F)F)N

Specifications

Melting Point -35°C to -33°C
Density 1.295
Boiling Point 67°C to 68°C (15 mmHg)
Flash Point 65°C (149°F)
Odor Recognizable
Refractive Index 1.48
Quantity 100 g
UN Number UN2942
Beilstein 879494
Formula Weight 161.13
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)aniline
Show More Show Less
Hazard Category H227-H302-H315-H317-H319-H373
Hazard Statement GHS H Statement
H318-H373-H302-H335-H227-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-TRIFLUOROMETHYLANILINE
EINECSNumber 201-806-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only