missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzyl Bromide 98.0+%, TCI America™

Catalog No. T23111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23111G 1 g
T23115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23111G Supplier TCI America Supplier No. T23111G
Only null left

Chemical Identifiers

CAS 198649-68-2
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.034
MDL Number MFCD00236321
InChI Key TUNSVUOTVLWNQT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzyl bromide, 1-bromomethyl-2-trifluoromethoxy benzene, 2-trifluoromethoxy benzylbromide, alpha-bromo-2-trifluoromethoxy toluene, 2-trifluoromethoxybenzyl bromide, 2-trifluoromethoxy-benzyl bromide, benzene, 1-bromomethyl-2-trifluoromethoxy, 2-bromomethyl phenoxy trifluoromethane, acmc-1c20t
PubChem CID 2777254
IUPAC Name 1-(bromomethyl)-2-(trifluoromethoxy)benzene
SMILES C1=CC=C(C(=C1)CBr)OC(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 255.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzyl Bromide