missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzenesulfonamide 98.0+%, TCI America™

Supplier:  TCI America T26045G

 View more versions of this product

Catalog No. T26045G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(Trifluoromethoxy)benzenesulfonamide
188°C
MFCD01320751
2-trifluoromethoxy benzenesulfonamide, benzenesulfonamide, 2-trifluoromethoxy, unii-3dq25354mb, 2-trifluoromethoxy benzenesulphonamide, 2-trifluoromethoxy benzene-1-sulfonamide, acmc-209itm, dsstox_cid_24930, dsstox_rid_80595, dsstox_gsid_44930, ksc222c3t
NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F
241.18
241.18
Crystalline Powder
37526-59-3
C7H6F3NO3S
5 g
HIFGQHGWMTZMOH-UHFFFAOYSA-N
2-(trifluoromethoxy)benzene-1-sulfonamide
2777208
≥98.0% (GC,T)

Chemical Identifiers

37526-59-3
241.18
HIFGQHGWMTZMOH-UHFFFAOYSA-N
2777208
NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F
C7H6F3NO3S
MFCD01320751
2-trifluoromethoxy benzenesulfonamide, benzenesulfonamide, 2-trifluoromethoxy, unii-3dq25354mb, 2-trifluoromethoxy benzenesulphonamide, 2-trifluoromethoxy benzene-1-sulfonamide, acmc-209itm, dsstox_cid_24930, dsstox_rid_80595, dsstox_gsid_44930, ksc222c3t
2-(trifluoromethoxy)benzene-1-sulfonamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions