missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethoxy)benzenesulfonamide 98.0+%, TCI America™
$42.87 - $123.03
Chemical Identifiers
| CAS | 37526-59-3 |
|---|---|
| Molecular Formula | C7H6F3NO3S |
| Molecular Weight (g/mol) | 241.18 |
| MDL Number | MFCD01320751 |
| InChI Key | HIFGQHGWMTZMOH-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethoxy benzenesulfonamide, benzenesulfonamide, 2-trifluoromethoxy, unii-3dq25354mb, 2-trifluoromethoxy benzenesulphonamide, 2-trifluoromethoxy benzene-1-sulfonamide, acmc-209itm, dsstox_cid_24930, dsstox_rid_80595, dsstox_gsid_44930, ksc222c3t |
| PubChem CID | 2777208 |
| IUPAC Name | 2-(trifluoromethoxy)benzene-1-sulfonamide |
| SMILES | NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F |
Chemical Identifiers
| 37526-59-3 | |
| 241.18 | |
| HIFGQHGWMTZMOH-UHFFFAOYSA-N | |
| 2777208 | |
| NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F |
| C7H6F3NO3S | |
| MFCD01320751 | |
| 2-trifluoromethoxy benzenesulfonamide, benzenesulfonamide, 2-trifluoromethoxy, unii-3dq25354mb, 2-trifluoromethoxy benzenesulphonamide, 2-trifluoromethoxy benzene-1-sulfonamide, acmc-209itm, dsstox_cid_24930, dsstox_rid_80595, dsstox_gsid_44930, ksc222c3t | |
| 2-(trifluoromethoxy)benzene-1-sulfonamide |
Specifications
| 37526-59-3 | |
| C7H6F3NO3S | |
| 5 g | |
| HIFGQHGWMTZMOH-UHFFFAOYSA-N | |
| 2-(trifluoromethoxy)benzene-1-sulfonamide | |
| 2777208 | |
| ≥98.0% (GC,T) | |
| 2-(Trifluoromethoxy)benzenesulfonamide |
| 188°C | |
| MFCD01320751 | |
| 2-trifluoromethoxy benzenesulfonamide, benzenesulfonamide, 2-trifluoromethoxy, unii-3dq25354mb, 2-trifluoromethoxy benzenesulphonamide, 2-trifluoromethoxy benzene-1-sulfonamide, acmc-209itm, dsstox_cid_24930, dsstox_rid_80595, dsstox_gsid_44930, ksc222c3t | |
| NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F | |
| 241.18 | |
| 241.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No