missing translation for 'onlineSavingsMsg'
Learn More

[2-(Tetrahydropyran-4-yloxy)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC52209DA

 View more versions of this product

Catalog No. CC52209DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

[2-(Tetrahydropyran-4-yloxy)phenyl]methanol
96.5
97%
C12H16O3
1g
RMRHXMQGVDIPCL-UHFFFAOYSA-N
[2-(oxan-4-yloxy)phenyl]methanol
22932192
97%
478189-93-4
100.0
Amber Glass Bottle
MFCD09064960
2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
C1COCCC1OC2=CC=CC=C2CO
208.257
208.26

Chemical Identifiers

478189-93-4
208.257
RMRHXMQGVDIPCL-UHFFFAOYSA-N
22932192
C1COCCC1OC2=CC=CC=C2CO
C12H16O3
MFCD09064960
2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
[2-(oxan-4-yloxy)phenyl]methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions