missing translation for 'onlineSavingsMsg'
Learn More

[2-(Tetrahydropyran-4-yloxy)phenyl]methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 478189-93-4
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD09064960
InChI Key RMRHXMQGVDIPCL-UHFFFAOYSA-N
Synonym 2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
PubChem CID 22932192
IUPAC Name [2-(oxan-4-yloxy)phenyl]methanol
SMILES C1COCCC1OC2=CC=CC=C2CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC52209DA
SDS View Documents
Thermo Scientific Chemicals
CC52209DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC52209DE
SDS View Documents
Thermo Scientific Chemicals
CC52209DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

478189-93-4
208.257
RMRHXMQGVDIPCL-UHFFFAOYSA-N
22932192
C1COCCC1OC2=CC=CC=C2CO
C12H16O3
MFCD09064960
2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
[2-(oxan-4-yloxy)phenyl]methanol

Specifications

478189-93-4
100.0
Amber Glass Bottle
MFCD09064960
2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
C1COCCC1OC2=CC=CC=C2CO
208.257
208.26
[2-(Tetrahydropyran-4-yloxy)phenyl]methanol
96.5
97%
C12H16O3
1g
RMRHXMQGVDIPCL-UHFFFAOYSA-N
[2-(oxan-4-yloxy)phenyl]methanol
22932192
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents