missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-p-cresol 98.0+%, TCI America™

Catalog No. B091025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B091025G 25 g
B0910500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B091025G Supplier TCI America Supplier No. B091025G

Chemical Identifiers

CAS 2409-55-4
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002381
InChI Key IKEHOXWJQXIQAG-UHFFFAOYSA-N
Synonym 2-tert-butyl-4-methylphenol, 2-tert-butyl-p-cresol, 2-t-butyl-p-cresol, p-cresol, 2-tert-butyl, o-tert-butyl-p-cresol, 2-t-butyl-4-methylphenol, 4-methyl-2-tert-butylphenol, 2-terc.butyl-p-kresol, phenol, 2-1,1-dimethylethyl-4-methyl, 4-methyl-6-t-butylphenol
PubChem CID 17004
IUPAC Name 2-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)C(C)(C)C

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 244°C
Quantity 25 g
UN Number 2430
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-tert-Butyl-p-cresol
EINECSNumber (3)-0521
RTECSNumber GO7000000
TSCA Yes