missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-methylphenol, 99%

Catalog No. AC107961000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107961000 100 g Glass bottle
AC107965000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107961000 Supplier Thermo Scientific Chemicals Supplier No. 107961000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2409-55-4 | C11H16O | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2409-55-4
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00002381
InChI Key IKEHOXWJQXIQAG-UHFFFAOYSA-N
Synonym 2-tert-butyl-4-methylphenol, 2-tert-butyl-p-cresol, 2-t-butyl-p-cresol, p-cresol, 2-tert-butyl, o-tert-butyl-p-cresol, 2-t-butyl-4-methylphenol, 4-methyl-2-tert-butylphenol, 2-terc.butyl-p-kresol, phenol, 2-1,1-dimethylethyl-4-methyl, 4-methyl-6-t-butylphenol
PubChem CID 17004
IUPAC Name 2-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 53.0°C
Color Beige-Yellow to White
Boiling Point 237.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H3(CH3)OH
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aceton, benzene, chloroform
Formula Weight 164.25
Percent Purity 99%
Physical Form Crystalline Mass
Chemical Name or Material 2-tert-Butyl-4-methylphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-314-6
RTECSNumber GO7000000
TSCA TSCA

RUO – Research Use Only