missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butyl)-4,6-dimethylphenol, 97%

Catalog No. p-7112909
Change view
Click to view available options
Quantity:
10 g, 10g
50 g, 50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462570100 10 g, 10g
AC462570500 50 g, 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462570100 Supplier Thermo Scientific Chemicals Supplier No. 462570100
Only null left

CAS: 1879-09-0 | C12H18O | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1879-09-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00002234
InChI Key OPLCSTZDXXUYDU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-tert-butylphenol, 2-tert-butyl-4,6-dimethylphenol, 6-tert-butyl-2,4-dimethylphenol, topanol a, 6-tert-butyl-2,4-xylenol, 6-t-butyl-2,4-xylenol, prodox 340, phenol, 2-1,1-dimethylethyl-4,6-dimethyl, 6-t-butyl-2,4-dimethylphenol, 2,4-xylenol, 6-tert-butyl
PubChem CID 15884
IUPAC Name 2-tert-butyl-4,6-dimethylphenol
SMILES CC1=CC(C)=C(O)C(=C1)C(C)(C)C

Specifications

Melting Point 22.0°C to 23.0°C
Boiling Point 249.0°C
Quantity 10 g, 10g
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2(OH)C6H2C(CH3)3
Formula Weight 178.27
Percent Purity 97%
Chemical Name or Material 2-(tert-Butyl)-4,6-dimethylphenol
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Toxic to aquatic
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Remove/Take off immediately all
EINECSNumber 217-533-1

RUO – Research Use Only