missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidone 98.0+%, TCI America™

Supplier:  TCI America P057525G

 View more versions of this product

Catalog No. P057525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Pyrrolidone
White-Yellow
C4H7NO
25 g
HNJBEVLQSNELDL-UHFFFAOYSA-N
pyrrolidin-2-one
12025
85.11
Crystalline Lumps
616-45-5
245°C
MFCD00005270
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
C1CC(=O)NC1
85.106
CHEBI:36592
≥98.0% (GC)

Chemical Identifiers

616-45-5
85.106
HNJBEVLQSNELDL-UHFFFAOYSA-N
12025
pyrrolidin-2-one
C4H7NO
MFCD00005270
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
CHEBI:36592
C1CC(=O)NC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0112

RTECSNumber : UY5715000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions