missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrrolidone 98.0+%, TCI America™
$29.83 - $68.97
Chemical Identifiers
| CAS | 616-45-5 |
|---|---|
| Molecular Formula | C4H7NO |
| Molecular Weight (g/mol) | 85.106 |
| MDL Number | MFCD00005270 |
| InChI Key | HNJBEVLQSNELDL-UHFFFAOYSA-N |
| Synonym | 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam |
| PubChem CID | 12025 |
| ChEBI | CHEBI:36592 |
| IUPAC Name | pyrrolidin-2-one |
| SMILES | C1CC(=O)NC1 |
Chemical Identifiers
| 616-45-5 | |
| 85.106 | |
| HNJBEVLQSNELDL-UHFFFAOYSA-N | |
| 12025 | |
| pyrrolidin-2-one |
| C4H7NO | |
| MFCD00005270 | |
| 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam | |
| CHEBI:36592 | |
| C1CC(=O)NC1 |
Specifications
| 616-45-5 | |
| 245°C | |
| MFCD00005270 | |
| 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam | |
| C1CC(=O)NC1 | |
| 85.106 | |
| CHEBI:36592 | |
| ≥98.0% (GC) | |
| 2-Pyrrolidone |
| White-Yellow | |
| C4H7NO | |
| 25 g | |
| HNJBEVLQSNELDL-UHFFFAOYSA-N | |
| pyrrolidin-2-one | |
| 12025 | |
| 85.11 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (5)-0112
RTECSNumber : UY5715000
TSCA : Yes