accessibility menu, dialog, popup

UserName

CAS RN 616-45-5

IUPAC Name: pyrrolidin-2-one
Synonyms: pyrrolidone 2-pyrol aminobutyric lactam 2-pyrrolidone-butyrolactam gamma-aminobutyric lactam 2-pyrrolidone, hydrotribromide gamma-aminobutyric acid lactam M-pyrol hydrobromide 2-pyrrolidone LAM
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: HNJBEVLQSNELDL-UHFFFAOYSA-N
SMILES: O=C1CCCN1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)