missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridylacetic Acid Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P107625G
Specifications
| 2-Pyridylacetic Acid Hydrochloride | |
| 130°C | |
| MFCD00012812 | |
| 2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride | |
| C1=CC=NC(=C1)CC(=O)O.Cl | |
| 173.596 | |
| 173.60 | |
| Crystalline Powder |
| 16179-97-8 | |
| C7H8ClNO2 | |
| 25 g | |
| MQVISALTZUNQSK-UHFFFAOYSA-N | |
| 2-pyridin-2-ylacetic acid;hydrochloride | |
| 85317 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 16179-97-8 | |
| 173.596 | |
| MQVISALTZUNQSK-UHFFFAOYSA-N | |
| 85317 | |
| C1=CC=NC(=C1)CC(=O)O.Cl |
| C7H8ClNO2 | |
| MFCD00012812 | |
| 2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride | |
| 2-pyridin-2-ylacetic acid;hydrochloride |
Safety and Handling
TSCA : Yes