missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridylacetic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134929
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10765G 5 g
P107625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10765G Supplier TCI America Supplier No. P10765G
Only null left

Chemical Identifiers

CAS 16179-97-8
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012812
InChI Key MQVISALTZUNQSK-UHFFFAOYSA-N
Synonym 2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride
PubChem CID 85317
IUPAC Name 2-pyridin-2-ylacetic acid;hydrochloride
SMILES C1=CC=NC(=C1)CC(=O)O.Cl

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 173.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyridylacetic Acid Hydrochloride