missing translation for 'onlineSavingsMsg'
Learn More

(2-Pyridyl)thiourea 98.0+%, TCI America™

Supplier:  TCI America P227225G

 View more versions of this product

Catalog No. P227225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(2-Pyridyl)thiourea
148°C
C6H7N3S
25 g
1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea
NC(=S)NC1=CC=CC=N1
153.20
153.20
Crystalline Powder
14294-11-2
White-Yellow
MFCD00041227
2811
SLUHLANJIVXTRQ-UHFFFAOYSA-N
(pyridin-2-yl)thiourea
1490491
≥98.0% (HPLC,T)

Chemical Identifiers

14294-11-2
153.20
SLUHLANJIVXTRQ-UHFFFAOYSA-N
1490491
NC(=S)NC1=CC=CC=N1
C6H7N3S
MFCD00041227
1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea
(pyridin-2-yl)thiourea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions