missing translation for 'onlineSavingsMsg'
Learn More

(2-Pyridyl)thiourea 98.0+%, TCI America™

Catalog No. p-7135487
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22725G 5 g
P227225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22725G Supplier TCI America Supplier No. P22725G

Chemical Identifiers

CAS 14294-11-2
Molecular Formula C6H7N3S
Molecular Weight (g/mol) 153.20
MDL Number MFCD00041227
InChI Key SLUHLANJIVXTRQ-UHFFFAOYSA-N
Synonym 1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea
PubChem CID 1490491
IUPAC Name (pyridin-2-yl)thiourea
SMILES NC(=S)NC1=CC=CC=N1

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 153.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Pyridyl)thiourea