missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propyltoluene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 185°C |
| Quantity | 5 mL |
| UN Number | 3295 |
| Formula Weight | 134.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Propyltoluene |
Safety and Handling
| EINECSNumber | (3)-0015 |
| TSCA | Yes |