accessibility menu, dialog, popup

UserName

CAS RN 1074-17-5

IUPAC Name: 1-methyl-2-propylbenzene
Synonyms: 1-methyl-2-propylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: YQZBFMJOASEONC-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results