missing translation for 'onlineSavingsMsg'
Learn More

2-Propyltoluene 98.0+%, TCI America™

Catalog No. P07475ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07475ML 5 mL
P074725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07475ML Supplier TCI America Supplier No. P07475ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1074-17-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00059246
InChI Key YQZBFMJOASEONC-UHFFFAOYSA-N
Synonym 1-Methyl-2-propylbenzene
PubChem CID 14091
IUPAC Name 1-methyl-2-propylbenzene
SMILES CCCC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 185°C
Quantity 5 mL
UN Number 3295
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propyltoluene
EINECSNumber (3)-0015
TSCA Yes