missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylthiazole 98.0+%, TCI America™

Supplier:  TCI America P224725G

 View more versions of this product

Catalog No. P224725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Propionylthiazole
Yellow
C6H7NOS
25 g
TYRAENAWSLPSLW-UHFFFAOYSA-N
1-(1,3-thiazol-2-yl)propan-1-one
65288
≥98.0% (GC)
43039-98-1
107°C
MFCD01681404
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
CCC(=O)C1=NC=CS1
141.188
141.19
Liquid

Chemical Identifiers

43039-98-1
141.188
TYRAENAWSLPSLW-UHFFFAOYSA-N
65288
CCC(=O)C1=NC=CS1
C6H7NOS
MFCD01681404
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
1-(1,3-thiazol-2-yl)propan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XJ5123000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions