missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propionylthiazole 98.0+%, TCI America™
$156.59 - $467.87
Chemical Identifiers
| CAS | 43039-98-1 |
|---|---|
| Molecular Formula | C6H7NOS |
| Molecular Weight (g/mol) | 141.188 |
| MDL Number | MFCD01681404 |
| InChI Key | TYRAENAWSLPSLW-UHFFFAOYSA-N |
| Synonym | 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k |
| PubChem CID | 65288 |
| IUPAC Name | 1-(1,3-thiazol-2-yl)propan-1-one |
| SMILES | CCC(=O)C1=NC=CS1 |
Chemical Identifiers
| 43039-98-1 | |
| 141.188 | |
| TYRAENAWSLPSLW-UHFFFAOYSA-N | |
| 65288 | |
| CCC(=O)C1=NC=CS1 |
| C6H7NOS | |
| MFCD01681404 | |
| 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
| 1-(1,3-thiazol-2-yl)propan-1-one |
Specifications
| 43039-98-1 | |
| 107°C | |
| MFCD01681404 | |
| 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
| CCC(=O)C1=NC=CS1 | |
| 141.188 | |
| 141.19 | |
| Liquid |
| Yellow | |
| C6H7NOS | |
| 5 g | |
| TYRAENAWSLPSLW-UHFFFAOYSA-N | |
| 1-(1,3-thiazol-2-yl)propan-1-one | |
| 65288 | |
| ≥98.0% (GC) | |
| 2-Propionylthiazole |
Safety and Handling
RTECSNumber : XJ5123000
TSCA : Yes