missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), Honeywell™ Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)
Supplier: Honeywell Chemicals 335394X25LGL
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Neutral | |
| -89.5°C | |
| Colorless | |
| 77°C | |
| 0.02 | |
| 750°F | |
| Glass Bottle | |
| n20/D 1.3770 to 1.3780 | |
| C3H8O | |
| MFCD00011674 | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.8% (GC) |
| 2-Propanol | |
| 67-63-0 | |
| 0.785g/mL at 25°C | |
| 4 x 2.5 L | |
| 12°C (53.6°F) | |
| Alcohol-like | |
| Liquid | |
| 0 to 0.001% | |
| (CH3)2CHOH | |
| 33mmHg (20°C), 44mmHg (25°C) | |
| 635639 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1g/mol | |
| ACS/Reagent ISO/Reagent Ph. Eur./puriss. p.a. |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00011674 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Safety and Handling
P210-P261-P305 + P351 + P338
DOTInformation : Class 3 - PG 2 - Isopropanol
RTECSNumber : 200-661-7