missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), Honeywell™ Riedel-de Haën™

Catalog No. p-7120081
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046273 1 L Glass Bottle
60046272 1 L Poly Bottle
60046275 2.5 L Glass Bottle
60046274 2.5 L Poly Bottle
60046279 5 L Poly Bottle
60046280 6 x 1 L Poly Bottle
60046278 4 x 2.5 L Glass Bottle
60046277 4 x 2.5 L Poly Bottle
60046270 10 L Stainless Steel Can
60046271 155 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 60046273 Supplier Honeywell Chemicals Supplier No. 335391LGL
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Neutral
Melting Point -89.5°C
Color Colorless
Density 0.785g/mL at 25°C
Boiling Point 77°C
Quantity 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.3770 to 1.3780
Residue after Evaporation 0 to 0.001%
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/Reagent ISO/Reagent Ph. Eur./puriss. p.a.
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
DOTInformation Class 3 - PG 2 - Isopropanol
RTECSNumber 200-661-7