missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), Honeywell™ Riedel-de Haën™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)

Supplier:  Honeywell Chemicals 335394X25L

 View more versions of this product

Catalog No. 60046277


Only null left
Explore available promotions

Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Specifications

Neutral
-89.5°C
Colorless
77°C
0.02
750°F
Poly Bottle
n20/D 1.3770 to 1.3780
C3H8O
MFCD00011674
UN1219
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.8% (GC)
2-Propanol
67-63-0
0.785g/mL at 25°C
4 x 2.5 L
12°C (53.6°F)
Alcohol-like
Liquid
0 to 0.001%
(CH3)2CHOH
33mmHg (20°C), 44mmHg (25°C)
635639
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1g/mol
ACS/Reagent ISO/Reagent Ph. Eur./puriss. p.a.

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

DOTInformation : Class 3 - PG 2 - Isopropanol

RTECSNumber : 200-661-7

SDS
Documents

Documents

Promotions

Promotions