missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, ≥99.9% (GC), MilliporeSigma™ Supelco™
2-Propanol is a secondary alcohol, which can find a wide number of applications as a solvent.
Supplier: Merck Emd Millipore 9123710MLF
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
| 67-63-0 | |
| 44 mmHg (25°C) | |
| 0.785 g/mL (at 25°C) | |
| Neat | |
| MFCD00011674 | |
| 82°C | |
| Completely soluble in water | |
| 10 mL | |
| ≥99.9% (GC) |
| 750°F | |
| 2.1 (vs. air) | |
| 12°C | |
| (CH3)2 CHOH | |
| -89.5°C | |
| sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol | |
| Analytical Standard | |
| 60.10 | |
| Analytical Standard |