missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-5-propylbenzene 98.0+%, TCI America™

Catalog No. p-7128669
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51341G 1 g
D51345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51341G Supplier TCI America Supplier No. D51341G

Chemical Identifiers

CAS 183245-00-3
Molecular Formula C9H10F2
Molecular Weight (g/mol) 156.18
MDL Number MFCD12405568
InChI Key AXTNQJKYTXEYRM-UHFFFAOYSA-N
PubChem CID 18979661
IUPAC Name 1,3-difluoro-5-propylbenzene
SMILES CCCC1=CC(F)=CC(F)=C1

Specifications

Color Colorless
Boiling Point 170°C
Quantity 1 g
UN Number 1993
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Difluoro-5-propylbenzene