accessibility menu, dialog, popup

UserName

CH3FF

CAS RN 183245-00-3

IUPAC Name: 1,3-difluoro-5-propylbenzene
Synonyms: 1,3-difluoro-5-propylbenzene
Molecular Weight (g/mol): 156.18
Molecular Formula: C9H10F2
InChi Key: AXTNQJKYTXEYRM-UHFFFAOYSA-N
SMILES: CCCC1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results