Learn More
2-Phenylthioacetamide, 97%
CAS: 645-54-5 | C8H9NS | 151.227 g/mol
Supplier: Thermo Scientific Chemicals H2651403
Description
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.
Solubility
Slightly Soluble in water (1.8 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 2-Phenylthioacetamide | |
| 91°C to 95°C | |
| MFCD00022177 | |
| 2-phenylthioacetamide, benzeneethanethioamide, phenylthioacetamide, acetamide, 2-phenylthio, benzeneethanimidothioic acid, phenylacetothioamide, 1-amino-2-phenylethane-1-thione, a-phenylthioacetamide, 2-phenyl-thioacetamide, alpha-phenylthioacetamide | |
| CJXBHFANXQMZBF-UHFFFAOYSA-N | |
| 2-phenylethanethioamide | |
| 731368 | |
| 97% |
| 645-54-5 | |
| C8H9NS | |
| 1 g | |
| Slightly Soluble in water (1.8g/L) (25°C). | |
| C1=CC=C(C=C1)CC(=S)N | |
| 151.227 | |
| 151.23 |
Chemical Identifiers
| 645-54-5 | |
| 151.227 | |
| CJXBHFANXQMZBF-UHFFFAOYSA-N | |
| 731368 | |
| C1=CC=C(C=C1)CC(=S)N |
| C8H9NS | |
| MFCD00022177 | |
| 2-phenylthioacetamide, benzeneethanethioamide, phenylthioacetamide, acetamide, 2-phenylthio, benzeneethanimidothioic acid, phenylacetothioamide, 1-amino-2-phenylethane-1-thione, a-phenylthioacetamide, 2-phenyl-thioacetamide, alpha-phenylthioacetamide | |
| 2-phenylethanethioamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only