missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylsulfonylmethyl)benzaldehyde 97.0+%, TCI America™

Catalog No. p14551g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14551G Supplier TCI America Supplier No. P14551G

Chemical Identifiers

CAS 468751-38-4
Molecular Formula C14H12O3S
Molecular Weight (g/mol) 260.31
MDL Number MFCD06797157
InChI Key KDTVRUZABJBZKA-UHFFFAOYSA-N
Synonym 2-(Benzenesulfonylmethyl)benzaldehyde, 2-Formylbenzyl Phenyl Sulfone
PubChem CID 11345986
IUPAC Name 2-[(benzenesulfonyl)methyl]benzaldehyde
SMILES O=CC1=CC=CC=C1CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 1 g
Formula Weight 260.31
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Phenylsulfonylmethyl)benzaldehyde