missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol 99.0+%, TCI America™

Catalog No. p-7134482
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020025G 25 g
P0200500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P020025G Supplier TCI America Supplier No. P020025G
Only null left

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
IUPAC Name [1,1'-biphenyl]-2-ol
SMILES OC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 59°C
Color White
pH 11.2 to 11.6 (1% H2O soln.)
Quantity 25 g
UN Number 3077
Formula Weight 170.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylphenol
EINECSNumber (4)-0019
RTECSNumber DV5775000
TSCA Yes