accessibility menu, dialog, popup

UserName

CAS RN 90-43-7

IUPAC Name: [1,1'-biphenyl]-2-ol
Synonyms: o-phenylphenol nectryl o-xenol tumescal 0PE dowicide 2-phenylphenol (substance) caswell no. 623AA dowicide 1 dowicide A invalon OP
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: LLEMOWNGBBNAJR-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)