missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol Sodium Salt Tetrahydrate 80.0+%, TCI America™

Catalog No. P020225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020225G 25 g
P0202500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P020225G Supplier TCI America Supplier No. P020225G

Chemical Identifiers

CAS 132-27-4
Molecular Formula C12H9NaO
Molecular Weight (g/mol) 192.193
MDL Number MFCD00002209
InChI Key KSQXVLVXUFHGJQ-UHFFFAOYSA-M
Synonym sodium 1,1'-biphenyl-2-olate, sodium 2-biphenylate, sodium o-phenylphenate, 2-phenylphenol sodium salt, natriphene, orphenol, bactrol, o-phenylphenol sodium, sodium o-phenylphenol, dowicide a
PubChem CID 23675735
ChEBI CHEBI:82371
IUPAC Name sodium;2-phenylphenolate
SMILES C1=CC=C(C=C1)C2=CC=CC=C2[O-].[Na+]

Specifications

Melting Point 78°C
Quantity 25 g
UN Number 1759
Formula Weight 192.19
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylphenol Sodium Salt Tetrahydrate
EINECSNumber (4)-0020
RTECSNumber DV7700000
TSCA Yes