missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethylamine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P0086100G
Specifications
| 2-Phenylethylamine Hydrochloride | |
| 224°C | |
| C8H12ClN | |
| 100 g | |
| SKHIBNDAFWIOPB-UHFFFAOYSA-N | |
| 2-phenylethanamine;hydrochloride | |
| 9075 | |
| ≥98.0% (HPLC,T) |
| 156-28-5 | |
| White-Yellow | |
| MFCD00012893 | |
| 2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d | |
| C1=CC=C(C=C1)CCN.Cl | |
| 157.641 | |
| 157.64 | |
| Crystalline Powder |
Chemical Identifiers
| 156-28-5 | |
| 157.641 | |
| SKHIBNDAFWIOPB-UHFFFAOYSA-N | |
| 9075 | |
| C1=CC=C(C=C1)CCN.Cl |
| C8H12ClN | |
| MFCD00012893 | |
| 2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d | |
| 2-phenylethanamine;hydrochloride |
Safety and Handling
RTECSNumber : SH7175000
TSCA : Yes