accessibility menu, dialog, popup

UserName

CAS RN 156-28-5

IUPAC Name: hydrogen 2-phenylethan-1-amine chloride
Synonyms: hydrogen 2-phenylethylamine chloride
Molecular Weight (g/mol): 157.64
Molecular Formula: C8H12ClN
InChi Key: SKHIBNDAFWIOPB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)