missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134418
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P008625G 25 g
P0086100G 100 g
P0086500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P008625G Supplier TCI America Supplier No. P008625G
Only null left

Chemical Identifiers

CAS 156-28-5
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
MDL Number MFCD00012893
InChI Key SKHIBNDAFWIOPB-UHFFFAOYSA-N
Synonym 2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d
PubChem CID 9075
IUPAC Name 2-phenylethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CCN.Cl

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 25 g
Formula Weight 157.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylethylamine Hydrochloride
RTECSNumber SH7175000
TSCA Yes