missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl Isovalerate 98.0+%, TCI America™
Supplier: TCI America P200925ML
Specifications
| 2-Phenylethyl Isovalerate | |
| Colorless | |
| C13H18O2 | |
| 25 mL | |
| JIMGVOCOYZFDKB-UHFFFAOYSA-N | |
| 2-phenylethyl 3-methylbutanoate | |
| 8792 | |
| ≥98.0% (GC) |
| 140-26-1 | |
| 268°C | |
| MFCD00044322 | |
| phenylethyl isovalerate, phenethyl isovalerate, 2-phenylethyl isovalerate, butanoic acid, 3-methyl-, 2-phenylethyl ester, isovaleric acid, phenethyl ester, phenylethyl 3-methylbutyrate, phenethyl isovalerianate, 2-phenylethyl 3-methylbutirate, benzylcarbinyl isovalerate, phenethyl 3-methylbutyrate | |
| CC(C)CC(=O)OCCC1=CC=CC=C1 | |
| 206.285 | |
| 206.29 | |
| Liquid |
Chemical Identifiers
| 140-26-1 | |
| 206.285 | |
| JIMGVOCOYZFDKB-UHFFFAOYSA-N | |
| 8792 | |
| CC(C)CC(=O)OCCC1=CC=CC=C1 |
| C13H18O2 | |
| MFCD00044322 | |
| phenylethyl isovalerate, phenethyl isovalerate, 2-phenylethyl isovalerate, butanoic acid, 3-methyl-, 2-phenylethyl ester, isovaleric acid, phenethyl ester, phenylethyl 3-methylbutyrate, phenethyl isovalerianate, 2-phenylethyl 3-methylbutirate, benzylcarbinyl isovalerate, phenethyl 3-methylbutyrate | |
| 2-phenylethyl 3-methylbutanoate |
Safety and Handling
EINECSNumber : (3)-1040&(3)-1045
RTECSNumber : NY1511500
TSCA : Yes