accessibility menu, dialog, popup

UserName

CAS RN 140-26-1

H3CCH3OO

CAS RN 140-26-1

IUPAC Name: 2-phenylethyl 3-methylbutanoate
Synonyms: phenethyl isovalerate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: JIMGVOCOYZFDKB-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)OCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results