missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl Isovalerate 98.0+%, TCI America™
$54.06 - $177.08
Chemical Identifiers
| CAS | 140-26-1 |
|---|---|
| Molecular Formula | C13H18O2 |
| Molecular Weight (g/mol) | 206.285 |
| MDL Number | MFCD00044322 |
| InChI Key | JIMGVOCOYZFDKB-UHFFFAOYSA-N |
| Synonym | phenylethyl isovalerate, phenethyl isovalerate, 2-phenylethyl isovalerate, butanoic acid, 3-methyl-, 2-phenylethyl ester, isovaleric acid, phenethyl ester, phenylethyl 3-methylbutyrate, phenethyl isovalerianate, 2-phenylethyl 3-methylbutirate, benzylcarbinyl isovalerate, phenethyl 3-methylbutyrate |
| PubChem CID | 8792 |
| IUPAC Name | 2-phenylethyl 3-methylbutanoate |
| SMILES | CC(C)CC(=O)OCCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P200925ML
|
TCI America
P200925ML |
25 mL |
Each for $54.06
|
|
|||||
|
P2009500ML
|
TCI America
P2009500ML |
500 mL |
Each for $177.08
|
|
|||||
Chemical Identifiers
| 140-26-1 | |
| 206.285 | |
| JIMGVOCOYZFDKB-UHFFFAOYSA-N | |
| 8792 | |
| CC(C)CC(=O)OCCC1=CC=CC=C1 |
| C13H18O2 | |
| MFCD00044322 | |
| phenylethyl isovalerate, phenethyl isovalerate, 2-phenylethyl isovalerate, butanoic acid, 3-methyl-, 2-phenylethyl ester, isovaleric acid, phenethyl ester, phenylethyl 3-methylbutyrate, phenethyl isovalerianate, 2-phenylethyl 3-methylbutirate, benzylcarbinyl isovalerate, phenethyl 3-methylbutyrate | |
| 2-phenylethyl 3-methylbutanoate |
Specifications
| 140-26-1 | |
| 268°C | |
| MFCD00044322 | |
| phenylethyl isovalerate, phenethyl isovalerate, 2-phenylethyl isovalerate, butanoic acid, 3-methyl-, 2-phenylethyl ester, isovaleric acid, phenethyl ester, phenylethyl 3-methylbutyrate, phenethyl isovalerianate, 2-phenylethyl 3-methylbutirate, benzylcarbinyl isovalerate, phenethyl 3-methylbutyrate | |
| CC(C)CC(=O)OCCC1=CC=CC=C1 | |
| 206.285 | |
| 206.29 | |
| Liquid |
| Colorless | |
| C13H18O2 | |
| 25 mL | |
| JIMGVOCOYZFDKB-UHFFFAOYSA-N | |
| 2-phenylethyl 3-methylbutanoate | |
| 8792 | |
| ≥98.0% (GC) | |
| 2-Phenylethyl Isovalerate |
Safety and Handling
EINECSNumber : (3)-1040&(3)-1045
RTECSNumber : NY1511500
TSCA : Yes