missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylbutyryl Chloride 97.0+%, TCI America™
Supplier: TCI America P166525G
Specifications
| 2-Phenylbutyryl Chloride | |
| Yellow | |
| C10H11ClO | |
| 25 g | |
| 2-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenylbutyrylchloride, butyryl chloride, 2-phenyl, acmc-209ipa, 2-phenyl butyryl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, .alpha.-phenylbutyryl chloride, 2-phenyl-2-ethylacetyl chloride | |
| CCC(C1=CC=CC=C1)C(=O)Cl | |
| 182.647 | |
| 182.65 | |
| Liquid |
| 36854-57-6 | |
| 125°C | |
| MFCD00018811 | |
| 3265 | |
| QGXMHCMPIAYMGT-UHFFFAOYSA-N | |
| 2-phenylbutanoyl chloride | |
| 98173 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 36854-57-6 | |
| 182.647 | |
| QGXMHCMPIAYMGT-UHFFFAOYSA-N | |
| 98173 | |
| CCC(C1=CC=CC=C1)C(=O)Cl |
| C10H11ClO | |
| MFCD00018811 | |
| 2-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenylbutyrylchloride, butyryl chloride, 2-phenyl, acmc-209ipa, 2-phenyl butyryl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, .alpha.-phenylbutyryl chloride, 2-phenyl-2-ethylacetyl chloride | |
| 2-phenylbutanoyl chloride |
Safety and Handling
EINECSNumber : (9)-1149
RTECSNumber : ET5957500
TSCA : No