accessibility menu, dialog, popup

UserName

CAS RN 36854-57-6

CH3ClO

CAS RN 36854-57-6

IUPAC Name: 2-phenylbutanoyl chloride
Synonyms: 2-phenylbutanoyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: QGXMHCMPIAYMGT-UHFFFAOYNA-N
SMILES: CCC(C(Cl)=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results