missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacrylic acid, 97%

Catalog No. p-7077772
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459050050 5 g
AC459050250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459050050 Supplier Thermo Scientific Chemicals Supplier No. 459050050
Only null left

CAS: 492-38-6 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-38-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
InChI Key ONPJWQSDZCGSQM-UHFFFAOYSA-N
Synonym 2-phenylacrylic acid, atropic acid, acrylic acid, 2-phenyl, 2-propenoic acid, 2-phenyl, unii-tw16ua35s0, benzeneacetic acid, .alpha.-methylene, .alpha.-phenyl acrylic acid, .alpha.-toluic acid, .alpha.-methylene, alpha-phenylacrylic acid, alpha-phenyl acrylic acid
PubChem CID 68114
IUPAC Name 2-phenylprop-2-enoic acid
SMILES C=C(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 104.0°C to 106.0°C
Boiling Point 267.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 2-Phenylacrylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wash face, hands and any exposed skin thoroughly after handling.
EINECSNumber 207-753-6

RUO – Research Use Only