missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacetamide 98.0+%, TCI America™

Catalog No. p-7134436
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P012025G 25 g
P0120500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P012025G Supplier TCI America Supplier No. P012025G
Only null left

Chemical Identifiers

CAS 103-81-1
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00059193
InChI Key LSBDFXRDZJMBSC-UHFFFAOYSA-N
Synonym benzeneacetamide, phenylacetamide, alpha-toluamide, alpha-phenylacetamide, phenylacetic acid amide, acetamide, 2-phenyl, phenyl-beta-acetylamine, alpha-toluimidic acid, 2-phenyl-acetamide, phenyl acetamide
PubChem CID 7680
ChEBI CHEBI:16562
IUPAC Name 2-phenylacetamide
SMILES NC(=O)CC1=CC=CC=C1

Specifications

Melting Point 156°C
Color White
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylacetamide
EINECSNumber (9)-1200
RTECSNumber AC7705000
TSCA Yes