missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol 98.0+%, TCI America™

Catalog No. p-7134491
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021325G 25 g
P0213100G 100 g
P0213500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P021325G Supplier TCI America Supplier No. P021325G
Only null left

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1

Specifications

Melting Point 32°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-2-propanol
EINECSNumber (3)-1013&(3)-1034
RTECSNumber DO4562000
TSCA Yes