missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-1,3-propanediol 98.0+%, TCI America™
Supplier: TCI America P12495G
Specifications
| 2-Phenyl-1,3-propanediol | |
| 52°C | |
| C9H12O2 | |
| 5 g | |
| BPBDZXFJDMJLIB-UHFFFAOYSA-N | |
| 2-phenylpropane-1,3-diol | |
| 254178 | |
| ≥98.0% (GC) |
| 1570-95-2 | |
| White-Yellow | |
| MFCD00236056 | |
| 2-phenyl-1,3-propanediol, 2-phenyl-1,3-propanediole, 2-phenyl-propane-1,3-diol, 2-phenyl-1,3-propane diol, unii-9f93674jbb, 1,3-propanediol, 2-phenyl, phenylpropanediol, pubchem20386, rarechem al bd 1370, acmc-1bz5a | |
| C1=CC=C(C=C1)C(CO)CO | |
| 152.193 | |
| 152.19 | |
| Crystalline Powder |
Chemical Identifiers
| 1570-95-2 | |
| 152.193 | |
| BPBDZXFJDMJLIB-UHFFFAOYSA-N | |
| 254178 | |
| C1=CC=C(C=C1)C(CO)CO |
| C9H12O2 | |
| MFCD00236056 | |
| 2-phenyl-1,3-propanediol, 2-phenyl-1,3-propanediole, 2-phenyl-propane-1,3-diol, 2-phenyl-1,3-propane diol, unii-9f93674jbb, 1,3-propanediol, 2-phenyl, phenylpropanediol, pubchem20386, rarechem al bd 1370, acmc-1bz5a | |
| 2-phenylpropane-1,3-diol |
Safety and Handling
TSCA : No