accessibility menu, dialog, popup

UserName

OHOH

CAS RN 1570-95-2

IUPAC Name: 2-phenylpropane-1,3-diol
Synonyms: 2-phenylpropane-1,3-diol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: BPBDZXFJDMJLIB-UHFFFAOYSA-N
SMILES: OCC(CO)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)