missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7135031
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12495G 5 g
P124925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12495G Supplier TCI America Supplier No. P12495G
Only null left

Chemical Identifiers

CAS 1570-95-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00236056
InChI Key BPBDZXFJDMJLIB-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-propanediol, 2-phenyl-1,3-propanediole, 2-phenyl-propane-1,3-diol, 2-phenyl-1,3-propane diol, unii-9f93674jbb, 1,3-propanediol, 2-phenyl, phenylpropanediol, pubchem20386, rarechem al bd 1370, acmc-1bz5a
PubChem CID 254178
IUPAC Name 2-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CO)CO

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-1,3-propanediol