missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl Isobutyrate 98.0+%, TCI America™

Catalog No. P182225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P182225G 25 g
P1822500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P182225G Supplier TCI America Supplier No. P182225G

Chemical Identifiers

CAS 103-60-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00027363
InChI Key MJTPMXWJHPOWGH-UHFFFAOYSA-N
Synonym Isobutyric Acid 2-Phenoxyethyl Ester
PubChem CID 61005
IUPAC Name 2-phenoxyethyl 2-methylpropanoate
SMILES CC(C)C(=O)OCCOC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 127°C
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethyl Isobutyrate
EINECSNumber (3)-0564
RTECSNumber UA2470910
TSCA Yes