accessibility menu, dialog, popup

UserName

H3CCH3OOO

CAS RN 103-60-6

IUPAC Name: 2-phenoxyethyl 2-methylpropanoate
Synonyms: phenoxyethyl isobutyrate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: MJTPMXWJHPOWGH-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OCCOC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results