missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

2-Phenoxyethanol is a compound, which belongs to the class of volatile organicshaving a faint rose like aromatic odor. It may find applications as a solvent in ball-point inks, resins and in organic synthesis. It may also be used in cosmetics as an antimicrobial agent and preservative.

$70.10 - $272.29

Specifications

CAS 122-99-6
Density 1.107 g/mL (at 20°C)
Flash Point 121°C
Physical Form Neat
Linear Formula C6H5 OCH2 CH2 OH
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012813
View Documents
Merck Emd Millipore
567531MLF
1 mL
Each for $70.10
Only null left
 
111012814
View Documents
Merck Emd Millipore
567535MLF
5 mL
Each for $272.29
Only null left
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

122-99-6
121°C
C6H5 OCH2 CH2 OH
11°C to 13°C
Ethylene glycol monophenyl ether; PHE; PHE-G; PHE-S; PhG; Phenylglycol
138.16
Analytical Standard
1.107 g/mL (at 20°C)
Neat
MFCD00002857
244°C to 246°C
Analytical Standard
≥99.5% (GC)
Videos
SDS
Documents

Documents